Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
clec3b
Entrez Gene ID
496907
Gene Name
C-type lectin domain family 3 member B
Gene Aliases
-
Chromosome Location
Chr.6: 63662247 - 63669801 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011424.1 NP_001011424.1 1 - 2 107 112
GenBank mRNA BC088053.1 - 1 - 2 107 112

View Full Product Details