Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cldn1
Entrez Gene ID
548421
Gene Name
claudin 1
Gene Aliases
claudin-1, cld1, ilvasc, semp1
Chromosome Location
Chr.5: 99661785 - 99673138 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015704.1 NP_001015704.1 1 - 2 370 88
GenBank mRNA BC089635.1 - 1 - 2 370 88

View Full Product Details