Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ctsl
Entrez Gene ID
548464
Gene Name
cathepsin L
Gene Aliases
catl, cpl-1, ctsl1, mep
Chromosome Location
Chr.3: 85602262 - 85607609 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015747.1 NP_001015747.1 3 - 4 465 72
XM_012966538.2 XP_012821992.1 3 - 4 369 72
GenBank mRNA BC089683.1 - 3 - 4 465 72

View Full Product Details