Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pglyrp1
Entrez Gene ID
548492
Gene Name
peptidoglycan recognition protein 1
Gene Aliases
pglyrp, pgrp, pgrp-s, pgrps, tag7, tnfsf3l
Chromosome Location
Chr.8: 49278187 - 49284275 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015775.1 NP_001015775.1 3 - 4 376 92
XM_012968332.2 XP_012823786.1 4 - 5 578 92
XM_012968333.2 XP_012823787.1 4 - 5 463 92
GenBank mRNA BC089715.1 - 3 - 4 376 92

View Full Product Details