Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
yrdc
Entrez Gene ID
548554
Gene Name
yrdC N6-threonylcarbamoyltransferase domain containing
Gene Aliases
drip3, irip, sua5
Chromosome Location
Chr.2: 57893500 - 57903443 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC090125.1 - 4 - 5 690 63

View Full Product Details