Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cdc37
Entrez Gene ID
548666
Gene Name
cell division cycle 37
Gene Aliases
-
Chromosome Location
Chr.3: 127114831 - 127124010 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015912.1 NP_001015912.1 7 - 8 959 93
GenBank mRNA BC135957.1 - 7 - 8 981 93
CR926424.1 - 7 - 8 959 93

View Full Product Details