Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kctd10
Entrez Gene ID
549312
Gene Name
potassium channel tetramerization domain containing 10
Gene Aliases
-
Chromosome Location
Chr.1: 138278166 - 138290390 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016558.1 NP_001016558.1 4 - 5 534 112
XM_012962508.2 XP_012817962.1 5 - 6 690 112
GenBank mRNA CR760702.1 - 4 - 5 534 112

View Full Product Details