Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rassf7
Entrez Gene ID
549317
Gene Name
Ras association (RalGDS/AF-6) domain family (N-terminal) member 7
Gene Aliases
hras1
Chromosome Location
Chr.4: 39516448 - 39566127 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016563.1 NP_001016563.1 4 - 5 1326 63
XM_012961110.2 XP_012816564.1 5 - 6 1430 63
XM_012961111.2 XP_012816565.1 4 - 5 1412 63
XM_012961112.2 XP_012816566.1 4 - 5 1349 63
XM_018093052.1 XP_017948541.1 4 - 5 1412 63
XM_018093053.1 XP_017948542.1 4 - 5 1340 63
GenBank mRNA CR760685.1 - 4 - 5 1326 63
CU075861.1 - 5 - 6 1378 63

View Full Product Details