Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rpl11
Entrez Gene ID
549394
Gene Name
ribosomal protein L11
Gene Aliases
-
Chromosome Location
Chr.2: 86640536 - 86648640 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016640.2 NP_001016640.1 3 - 4 397 97
XM_012956992.1 XP_012812446.1 4 - 5 468 97
GenBank mRNA BC135522.1 - 3 - 4 410 97
CR760409.2 - 3 - 4 397 97

View Full Product Details