Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pdcd5
Entrez Gene ID
549765
Gene Name
programmed cell death 5
Gene Aliases
tfar19
Chromosome Location
Chr.4: 21328611 - 21336762 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017011.1 NP_001017011.1 3 - 4 172 103
GenBank mRNA CR762246.1 - 3 - 4 172 103

View Full Product Details