Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
acer2
Entrez Gene ID
549870
Gene Name
alkaline ceramidase 2
Gene Aliases
asah3l
Chromosome Location
Chr.1: 106911254 - 106932653 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017116.2 NP_001017116.1 5 - 6 795 80
XM_012963315.2 XP_012818769.1 4 - 5 984 80
XM_012963318.2 XP_012818772.1 5 - 6 790 80
GenBank mRNA BC168538.1 - 4 - 5 874 80
CR761183.2 - 5 - 6 795 80

View Full Product Details