Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
snx7
Entrez Gene ID
549959
Gene Name
sorting nexin 7
Gene Aliases
-
Chromosome Location
Chr.4: 92875491 - 92903939 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017205.2 NP_001017205.1 1 - 2 190 91
GenBank mRNA BC135154.1 - 1 - 2 201 91
CR760502.2 - 1 - 2 190 91

View Full Product Details