Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dda1
Entrez Gene ID
549970
Gene Name
DET1 and DDB1 associated 1
Gene Aliases
pcia1
Chromosome Location
Chr.1: 101218883 - 101227207 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017216.2 NP_001017216.1 2 - 3 148 132
GenBank mRNA BC090578.1 - 2 - 3 148 132
CR760393.2 - 1 - 2 87 132

View Full Product Details