Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
crybb1
Entrez Gene ID
550022
Gene Name
crystallin beta B1
Gene Aliases
catcn3
Chromosome Location
Chr.1: 140279832 - 140287480 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017268.3 NP_001017268.1 5 - 6 568 68
GenBank mRNA BX703001.1 - 5 - 6 568 68
CR855810.2 - 4 - 5 520 68
EL868269.1 - 1 - 2 292 68

View Full Product Details