Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kctd5
Entrez Gene ID
550048
Gene Name
potassium channel tetramerization domain containing 5
Gene Aliases
-
Chromosome Location
Chr.9: 22947117 - 22984496 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017294.2 NP_001017294.1 - 315 115
GenBank mRNA BC158374.1 - 2 - 3 319 115
CR855640.2 - - 315 115

View Full Product Details