Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
alkbh8
Entrez Gene ID
550051
Gene Name
alkB homolog 8, tRNA methyltransferase
Gene Aliases
-
Chromosome Location
Chr.2: 157051169 - 157072438 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017297.3 NP_001017297.2 9 - 10 1305 84
XM_012957058.2 XP_012812512.1 10 - 11 1420 84
XM_012957059.2 XP_012812513.1 10 - 11 1411 84
XM_012957060.2 XP_012812514.1 10 - 11 1431 84
XM_012957061.2 XP_012812515.1 10 - 11 1440 84
GenBank mRNA CR855607.2 - 10 - 11 1330 84

View Full Product Details