Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
b3gat1l
Entrez Gene ID
594896
Gene Name
beta-1,3-glucuronyltransferase 1 like
Gene Aliases
b3gat1, cd57, glcatp, glcuatp, hnk-1, hnk1, leu7, nk-1, nk1
Chromosome Location
Chr.2: 2203541 - 2265573 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030350.1 NP_001025521.1 1 - 2 114 75
GenBank mRNA AJ890838.1 - 1 - 2 114 75

View Full Product Details