Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
tpm2
Entrez Gene ID
594975
Gene Name
tropomyosin 2
Gene Aliases
AMCD1, DA1, DA2B, TMSB, beta-TMad, beta-TMemb, nem4, tpm2-a, tpm2-b
Chromosome Location
Chr.Un|NW_016683989: 4697 - 26312 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030416.1 NP_001025587.1 4 - 5 630 81
GenBank mRNA BC091033.1 - 4 - 5 630 81

View Full Product Details