Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
omg
Entrez Gene ID
594982
Gene Name
oligodendrocyte myelin glycoprotein
Gene Aliases
omgp
Chromosome Location
Chr.2: 101093052 - 101094523 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030423.1 NP_001025594.1 1 - 1 852 97
GenBank mRNA BC091043.1 - 1 - 1 852 97

View Full Product Details