Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dpt
Entrez Gene ID
594987
Gene Name
dermatopontin
Gene Aliases
-
Chromosome Location
Chr.2: 68364300 - 68390620 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030428.1 NP_001025599.1 1 - 2 308 81
GenBank mRNA BC091048.1 - 1 - 2 308 81

View Full Product Details