Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
matn1
Entrez Gene ID
595001
Gene Name
matrilin 1
Gene Aliases
cmp, crtm, xmatn1
Chromosome Location
Chr.2: 57369574 - 57379851 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030442.1 NP_001025613.1 1 - 2 100 74
XM_018091297.1 XP_017946786.1 1 - 2 133 74
GenBank mRNA BC091071.1 - 1 - 2 100 74

View Full Product Details