Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pglyrp1-like.1
Entrez Gene ID
595014
Gene Name
pglyrp1 protein
Gene Aliases
pglyrp1
Chromosome Location
Chr.8: 50316585 - 50379556 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030455.1 NP_001025626.1 3 - 4 412 60
XM_012968516.2 XP_012823970.1 5 - 6 601 60
GenBank mRNA BC091103.1 - 3 - 4 412 60

View Full Product Details