Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nup58
Entrez Gene ID
595038
Gene Name
nucleoporin 58kDa
Gene Aliases
nupl1, pro2463
Chromosome Location
Chr.2: 153352377 - 153368176 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030479.1 NP_001025650.1 1 - 2 172 81
GenBank mRNA BC091643.1 - 1 - 2 172 81
CR942452.2 - 1 - 2 179 81

View Full Product Details