Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
inha
Entrez Gene ID
613114
Gene Name
inhibin subunit alpha
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683550: 207132 - 209435 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032351.1 NP_001027522.1 1 - 2 454 81
GenBank mRNA BC098081.1 - 1 - 2 454 81

View Full Product Details