Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rala
Entrez Gene ID
548669
Gene Name
v-ral simian leukemia viral oncogene homolog A (ras related)
Gene Aliases
RAL-A, c-ral, ral
Chromosome Location
Chr.6: 81750395 - 81780840 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015915.2 NP_001015915.1 4 - 5 608 100
GenBank mRNA BC170836.1 - 4 - 5 595 100
BC170838.1 - 4 - 5 595 100
CR926418.2 - 4 - 5 608 100

View Full Product Details