Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
entpd7
Entrez Gene ID
548672
Gene Name
ectonucleoside triphosphate diphosphohydrolase 7
Gene Aliases
LALP1, NTPDase7
Chromosome Location
Chr.7: 27137843 - 27156935 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015918.2 NP_001015918.1 7 - 8 862 64
XM_012966551.2 XP_012822005.1 7 - 8 899 64
GenBank mRNA BC170622.1 - 6 - 7 825 64
CR926412.2 - 7 - 8 862 64
DQ118412.1 - 5 - 6 710 64

View Full Product Details