Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sap18
Entrez Gene ID
548920
Gene Name
Sin3A associated protein 18kDa
Gene Aliases
2hor0202, sap18p
Chromosome Location
Chr.2: 154700160 - 154703777 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016166.2 NP_001016166.1 1 - 2 152 76
GenBank mRNA BC080173.1 - 1 - 2 154 76
BC135392.1 - 1 - 2 136 76
CR760055.2 - 1 - 2 96 76
CR761908.2 - 1 - 2 152 76

View Full Product Details