Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnz1
Entrez Gene ID
548972
Gene Name
DNZDHHC/NEW1 zinc finger protein 11
Gene Aliases
-
Chromosome Location
Chr.8: 117360436 - 117362845 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016218.2 NP_001016218.1 - 421 95
GenBank mRNA BC171291.1 - - 421 95
BC171293.1 - - 421 95
CR926314.2 - - 421 95

View Full Product Details