Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
polr2d
Entrez Gene ID
549002
Gene Name
polymerase (RNA) II (DNA directed) polypeptide D
Gene Aliases
-
Chromosome Location
Chr.5: 121806868 - 121826506 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016248.2 NP_001016248.1 3 - 4 370 67
GenBank mRNA BC167273.1 - 3 - 4 660 67
BC167274.1 - 3 - 4 660 67
CR761767.2 - 3 - 4 370 67

View Full Product Details