Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
fam72a
Entrez Gene ID
549108
Gene Name
family with sequence similarity 72 member A
Gene Aliases
-
Chromosome Location
Chr.2: 65435034 - 65448907 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016354.2 NP_001016354.1 2 - 3 214 74
XM_012962201.2 XP_012817655.1 2 - 3 369 74
GenBank mRNA BC157357.1 - 1 - 2 135 74
BC170689.1 - 2 - 3 212 74
BC170693.1 - 2 - 3 212 74
CR761319.2 - 2 - 3 214 74

View Full Product Details