Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
psma2
Entrez Gene ID
549136
Gene Name
proteasome subunit alpha 2
Gene Aliases
XC3
Chromosome Location
Chr.6: 66009125 - 66017519 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016382.2 NP_001016382.1 7 - 8 600 65
GenBank mRNA BC157163.1 - 7 - 8 714 65
CR761217.2 - 7 - 8 600 65

View Full Product Details