Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
atxn3
Entrez Gene ID
549143
Gene Name
ataxin 3
Gene Aliases
at3, atx3, jos, mjd, mjd1, sca3
Chromosome Location
Chr.8: 66250732 - 66266063 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016389.2 NP_001016389.1 5 - 6 398 73
XM_012968399.2 XP_012823853.1 5 - 6 478 73
XM_012968400.2 XP_012823854.1 5 - 6 461 73
XM_012968401.2 XP_012823855.1 5 - 6 474 73
XM_012968402.2 XP_012823856.1 5 - 6 478 73
XM_012968403.2 XP_012823857.1 5 - 6 478 73
XM_012968404.2 XP_012823858.1 5 - 6 474 73
XM_012968405.2 XP_012823859.1 5 - 6 478 73
XM_018096253.1 XP_017951742.1 5 - 6 436 73
GenBank mRNA BC090121.1 - 4 - 5 382 73
BC157299.1 - 4 - 5 382 73
BC158945.1 - 5 - 6 479 73
CR761189.2 - 5 - 6 398 73

View Full Product Details