Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bet1
Entrez Gene ID
549196
Gene Name
Bet1 golgi vesicular membrane trafficking protein
Gene Aliases
-
Chromosome Location
Chr.6: 24655181 - 24659809 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016442.2 NP_001016442.1 1 - 2 197 112
GenBank mRNA BC167323.1 - 1 - 2 189 112
BC170583.1 - 1 - 2 195 112
BC170611.1 - 1 - 2 195 112
CR761023.2 - 1 - 2 197 112

Gene Symbol
LOC101730635
Entrez Gene ID
101730635
Gene Name
BET1 homolog
Gene Aliases
-
Chromosome Location
Chr.6: 24663358 - 24668208 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004919954.2 XP_004920011.1 1 - 2 262 112

View Full Product Details