Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
tekt3
Entrez Gene ID
549229
Gene Name
tektin 3
Gene Aliases
-
Chromosome Location
Chr.10: 39305053 - 39322689 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016475.2 NP_001016475.1 4 - 5 814 67
GenBank mRNA BC135257.1 - 4 - 5 818 67
CR760902.2 - 4 - 5 814 67

View Full Product Details