Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tex30
Entrez Gene ID
549282
Gene Name
testis expressed 30
Gene Aliases
c13orf27
Chromosome Location
Chr.2: 116023290 - 116030098 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016528.2 NP_001016528.1 4 - 5 526 84
XM_012956974.2 XP_012812428.1 4 - 5 590 84
XM_012956975.2 XP_012812429.1 4 - 5 485 84
GenBank mRNA BC170894.1 - 4 - 5 515 84
BC170896.1 - 4 - 5 515 84
CR926187.2 - 4 - 5 526 84

View Full Product Details