Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tekt1
Entrez Gene ID
549406
Gene Name
tektin 1
Gene Aliases
-
Chromosome Location
Chr.2: 36995722 - 37002721 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016652.2 NP_001016652.1 4 - 5 547 82
GenBank mRNA BC118772.1 - 4 - 5 571 82
CR760343.2 - 4 - 5 547 82

View Full Product Details