Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
c9orf85
Entrez Gene ID
549435
Gene Name
chromosome 9 open reading frame 85
Gene Aliases
-
Chromosome Location
Chr.1: 118197856 - 118210979 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016681.2 NP_001016681.1 2 - 3 224 98
GenBank mRNA CR760206.2 - 2 - 3 224 98

View Full Product Details