Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
edf1
Entrez Gene ID
549440
Gene Name
endothelial differentiation related factor 1
Gene Aliases
edf-1, mbf1
Chromosome Location
Chr.8: 369258 - 374488 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016686.2 NP_001016686.1 4 - 5 401 107
GenBank mRNA CR760196.2 - 4 - 5 401 107

View Full Product Details