Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
arl4c
Entrez Gene ID
549461
Gene Name
ADP ribosylation factor like GTPase 4C
Gene Aliases
arl7
Chromosome Location
Chr.9: 54280764 - 54283489 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016707.2 NP_001016707.1 1 - 1 655 66
GenBank mRNA CR760129.2 - 1 - 1 655 66

View Full Product Details