Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vamp3
Entrez Gene ID
549726
Gene Name
vesicle associated membrane protein 3
Gene Aliases
ceb, vamp-3
Chromosome Location
Chr.7: 93023876 - 93058713 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016972.2 NP_001016972.1 3 - 4 288 96
GenBank mRNA BC157185.1 - 3 - 4 303 96
CR848429.2 - 3 - 4 288 96

View Full Product Details