Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tspan3
Entrez Gene ID
549777
Gene Name
tetraspanin 3
Gene Aliases
tm4-a, tm4sf8, tspan-3
Chromosome Location
Chr.3: 80885785 - 80897113 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017023.2 NP_001017023.1 5 - 6 586 70
XM_018092102.1 XP_017947591.1 4 - 5 538 70
GenBank mRNA BC167324.1 - 5 - 6 684 70
CR762087.2 - 5 - 6 586 70

View Full Product Details