Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mapk12
Entrez Gene ID
549834
Gene Name
mitogen-activated protein kinase 12
Gene Aliases
Xp38gamma, erk3, erk6, p38gamma, prkm12, sapk-3, sapk3
Chromosome Location
Chr.3: 62040961 - 62094107 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017080.2 NP_001017080.1 5 - 6 824 101
XM_018092105.1 XP_017947594.1 6 - 7 1007 101
GenBank mRNA BC131830.1 - 6 - 7 850 101
BC167997.1 - 5 - 6 768 101
CR761485.2 - 5 - 6 824 101

View Full Product Details