Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ak6
Entrez Gene ID
549921
Gene Name
adenylate kinase 6
Gene Aliases
ad-004, cgi-137, cinap, cip, taf2g, taf9, tafii31, tafii32, tafiid32
Chromosome Location
Chr.1: 167447599 - 167457845 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017167.2 NP_001017167.1 2 - 3 137 104
GenBank mRNA CR760786.2 - 2 - 3 137 104

View Full Product Details