Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
marveld2
Entrez Gene ID
550046
Gene Name
MARVEL domain containing 2
Gene Aliases
dfnb49, marvd2, mrvldc2, tric, tricellulin
Chromosome Location
Chr.1: 167430070 - 167442736 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017292.2 NP_001017292.1 3 - 4 1422 104
GenBank mRNA BC123026.1 - 3 - 4 1412 104
CR855651.2 - 3 - 4 1422 104
CU075633.1 - 3 - 4 1472 104

View Full Product Details