Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lsm12
Entrez Gene ID
394627
Gene Name
LSM12 homolog
Gene Aliases
-
Chromosome Location
Chr.10: 14052158 - 14062045 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037267.1 NP_001032344.1 3 - 4 399 87
XM_012971597.2 XP_012827051.1 3 - 4 491 86
GenBank mRNA BC061398.1 - 3 - 4 401 86
BC091003.1 - 3 - 4 399 87

View Full Product Details