Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mhc1b2
Entrez Gene ID
447976
Gene Name
major histocompatibility complex nonclassical class Ib-2
Gene Aliases
ncib
Chromosome Location
Chr.8: 112653269 - 112667469 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037273.1 NP_001032350.2 2 - 3 327 100
NM_001247995.1 NP_001234924.1 2 - 3 328 100
GenBank mRNA BC075337.1 - 2 - 3 328 100
BC089748.1 - 2 - 3 327 100

View Full Product Details