Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mhc1b2
Entrez Gene ID
447976
Gene Name
major histocompatibility complex nonclassical class Ib-2
Gene Aliases
ncib
Chromosome Location
Chr.8: 112653269 - 112667469 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037273.1 NP_001032350.2 3 - 4 612 81
NM_001247995.1 NP_001234924.1 3 - 4 613 81
GenBank mRNA BC075337.1 - 3 - 4 613 81
BC089748.1 - 3 - 4 612 81

View Full Product Details