Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tmx4
Entrez Gene ID
496882
Gene Name
thioredoxin-related transmembrane protein 4
Gene Aliases
txndc13
Chromosome Location
Chr.5: 21640608 - 21672612 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039737.1 NP_001034826.1 1 - 2 332 106
GenBank mRNA BC088774.1 - 1 - 2 177 106
BC118714.1 - 1 - 2 220 106
CR761337.2 - 1 - 2 332 106

View Full Product Details