Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc5a8
Entrez Gene ID
394505
Gene Name
solute carrier family 5 member 8
Gene Aliases
ait, smct1, vito
Chromosome Location
Chr.3: 39204977 - 39226793 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA CR761796.2 - 4 - 5 731 77
CU075779.1 - 4 - 5 733 77

View Full Product Details