Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
enpp4
Entrez Gene ID
550082
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 4
Gene Aliases
E-NPP 4
Chromosome Location
Chr.5: 142898732 - 142908820 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017328.4 NP_001017328.2 2 - 3 1007 70
GenBank mRNA BC121205.1 - 2 - 3 1007 70
CR848578.2 - 2 - 3 955 70

View Full Product Details